Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDVAESPERD PHSPEDEEQP QGLSDDDILR DSGSDQDLDG AGVRASDLED EESAARGPSQ EEEDNHSDEE DRASEPKSQD QDSEVNELSR GPTSSPCEEE GDEGEEDRTS DLRDEASSVT RELDEHELDY DEEVPEEPAP AVQEDEAEKA GAEDDEEKGE GTPREEGKAG VQSVGEKESL EAAKEKKKED DDGEIDDGEI DDDDLEEGEV KDPSDRKVRP RPTCRFFMKG NCTWGMNCRF IHPGVNDKGN YSLITKADPF PPNGAPPLGP HPLMPANPWG GPVVDEILPP PPPEPPTESA WERGLRHAKE VLKKATIRKE QEPDFEEKRF TVTIGEDERE FDKENEVFRD WNSRIPRDVR DTVLEPYADP YYDYEIERFW RGGQYENFRV QYTETEPYHN YRERERERER ENRQRERERE RERDRERERR QREREREREH ERDKERQRRK EEWERERAKR DEKDRQHRDR DREKEREKEK GKPKPRSPQP PSRQAEPPKK EAATTGPQVK RADEWKDPWR RSKSPKKKLG VSVSPSRARR RRKTSASSAS ASNSSRSSSR SSSYSGSGSS RSRSRSSSYS SYSSRSSRHS SFSGSRSRSR SFSSSPSPSP TPSPHRPSIR TKGEPAPPPG KAGEKSVKKP APPPAPPQAT KTTAPVPEPT KPGDPREARR KERPARTPPR RRTLSGSGSG SGSSYSGSSS RSRSLSVSSV SSVSSATSSS SSAHSVDSED MYADLASPVS SASSRSPAPA QTRKEKGKSK KEDGVKEEKR KRDSSTQPPK SAKPPAGGKS SQQPSTPQQA PPGQPQQGTF VAHKEIKLTL LNKAADKGSR KRYEPSDKDR QSPPPAKRPN TSPDRGSRDR KSGGRLGSPK PERQRGQNSK APAAPADRKR QLSPQSKSSS KVTSVPGKAS DPGAASTKSG KASTLSRREE LLKQLKAVED AIARKRAKIP GKA
ZC3H18 contains 1 C3H1-type zinc finger.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: conserved nuclear protein NHN1; Nuclear protein NHN1; Zinc finger CCCH domain-containing protein 18
Gene Aliases: NHN1; ZC3H18
UniProt ID: (Human) Q86VM9
Entrez Gene ID: (Human) 124245
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support