Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that contains a tetratricopeptide repeat domain. The encoded protein also interacts with the rotavirus non-structural protein NSP3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Rotavirus 'X' associated non-structural protein; Rotavirus 'X'-associated non-structural protein; rotavirus X protein associated with NSP3; RoXaN; scrapie responsive gene 3; ubiquitous tetratricopeptide containing protein RoXaN; Zinc finger CCCH domain-containing protein 7B; zinc finger CCCH-type containing 7B
Gene Aliases: Gm627; KIAA1031; mKIAA1031; RoXaN; Scrg3; Zc2h7b; ZC3H7B
UniProt ID: (Human) Q9UGR2
Entrez Gene ID: (Human) 23264, (Mouse) 20286
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support