Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAEPTVCSF LTKVLCAHGG RMFLKDLRGH VELSEARLRD VLQRAGPERF LLQEVETQEG LGDAEAEAAA GAVGGGGTSA WRVVAVSSVR LCARYQRGEC QACDQLHFCR RHMLGKCPNR DCWSTCTLSH DIHTPVNMQV LKSHGLFGLN ENQLRILLLQ NDPCLLPEVC LLYNKGEALY GYCNLKDKCN KFHVCKSFVK GECKLQTCKR SHQLIHAASL KLLQDQGLNI PSVVNFQIIS TYKHIKLHKM LENTDNSSPS TEHSQGLEKQ GVHAAGAAEA GPLASVPAQS AKKPCPVSCE K (1-300 aa encoded by BC008842)
ZC3HAV1L is a protein coding gene. Gene ontology (GO) annotation include cytosol.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger CCCH-type antiviral protein 1-like; zinc finger CCCH-type, antiviral 1-like
Gene Aliases: C7orf39; ZC3HAV1L
UniProt ID: (Human) Q96H79
Entrez Gene ID: (Human) 92092
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support