Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAEPTVCSFL TKVLCAHGGR MFLKDLRGHV ELSEARLRDV LQRAGPERFL LQEVETQEGL GDAEAEAAAG AVGGGGTSAW RVVAVSSVRL CARYQRGECQ ACDQLHFCRR HMLGKCPNRD CWSTCTLSHD IHTPVNMQVL KSHGLFGLNE NQLRILLLQN DPCLLPEVCL LYNKGEALYG YCNLKDKCNK FHVCKSFVKG ECKLQTCKRS HQLIHAASLK LLQDQGLNIP SVVNFQIIST YKHIKLHKML ENTDNSSPST EHSQGLEKQG VHAAGAAEAG PLASVPAQSA KKPCPVSCEK
ZC3HAV1L is a protein coding gene. Gene ontology (GO) annotation include cytosol.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger CCCH-type antiviral protein 1-like; zinc finger CCCH-type, antiviral 1-like
Gene Aliases: C7orf39; ZC3HAV1L
UniProt ID: (Human) Q96H79
Entrez Gene ID: (Human) 92092
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support