Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MADEQEIMCK LESIKEIRNK TLQMEKIKAR LKAEFEALES EERHLKEYKQ EMDLLLQEKM AHVEELRLIH ADINVMENTI KQSENDLNKL LESTRRLHDE YKPLKEHVDA LRMTLGLQRL PDLCEEEEKL SLDYFEKQKA EWQTEPQEPP IPESLAAAAA AAQQLQVARK QDTRQTATFR QQPPPMKACL SCHQQIHRNA PICPLCKAKS RSRNPKKPKR KQDE
May play a role in neuronal development and in neuromuscular junction formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hepatocellular carcinoma-associated antigen 127; Zinc finger C4H2 domain-containing protein; zinc finger, C4H2 domain containing
Gene Aliases: HCA127; KIAA1166; WRWF; WWS; ZC4H2
UniProt ID: (Human) Q9NQZ6
Entrez Gene ID: (Human) 55906
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support