Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEESKTLKS ENHEPKKNVI CEESKAVQVI GNQTLKARND KSVKEIENSS PNRNSSKKNK QNDICIEKTE VKSCKVNAAN LPGPKDLGLV LRDQSHCKAK KFPNSPVKAE KATISQAKSE KATSLQAIAE KSPKSPNSVK AEKASSYQMK SEKVPSSPAE AEKGPSLLLK DMRQKTELQQ IGKKIPSSFT SVDKVNIEAV GGEKCALQNS PRSQKQQTCT DNTGDSDDSA SGIEDVSDDL SKMKNDESNK ENSSEMDYLE NATVIDESAL TPEQRL (1-275 aa encoded by B C048301)
WDR77 is a component of the 20S PRMT5-containing methyltransferase complex, which modifies specific arginines to dimethylarginines in several spliceosomal Sm proteins. This modification targets Sm proteins to the survival of motor neurons (SMN) complex for assembly into small nuclear ribonucleoprotein core particles (Friesen et al., 2002).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PAP associated domain containing 3; RP23-406B13.1; Terminal uridylyltransferase 4; TUTase 4; Zinc finger CCHC domain-containing protein 11; zinc finger, CCHC domain containing 11
Gene Aliases: KIAA0191; PAPD3; TUT4; ZCCHC11
UniProt ID: (Human) Q5TAX3
Entrez Gene ID: (Human) 23318
Molecular Function:
nucleotidyltransferase
transferase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support