Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGDTAKPYF VKRTKDRGTM DDDDFRRGHP QQDYLIIDDH AKGHGSKMEK GLQKKKITPG NYGNTPRKGP CAVSSNPYAF KNPIYSQPAW MNDSHKDQSK RWLSDEHTGN SDNWREFKPG PRIPVINRQR KDSFQENEDG YRWQDTRGCR TVRRLFHKDL TSLETTSEME AGSPENKKQR SRPRKPRKTR NEENEQDGDL EGPVIDESVL STKELLGLQQ AEERLKRDCI DRLKRRPRNY PTAKYTCRLC DV (1-251 aa encoded by BC032456)
Uridylyltransferase that mediates the terminal uridylation of mRNAs with short (less than 25 nucleotides) poly(A) tails, hence facilitating global mRNA decay (PubMed:19703396, PubMed:25480299). Essential for both oocyte maturation and fertility. Through 3' terminal uridylation of mRNA, sculpts, with TUT7, the maternal transcriptome by eliminating transcripts during oocyte growth. Involved in microRNA (miRNA)-induced gene silencing through uridylation of deadenylated miRNA targets (PubMed:25480299). Also functions as an integral regulator of microRNA biogenesiS using 3 different uridylation mechanisms (PubMed:25979828). Acts as a suppressor of miRNA biogenesis by mediating the terminal uridylation of some miRNA precursors, including that of let-7 (pre-let-7). Uridylated pre-let-7 RNA is not processed by Dicer and undergo degradation. Pre-let-7 uridylation is strongly enhanced in the presence of LIN28A (PubMed:22898984). In the absence of LIN28A, TUT7 and TUT4 monouridylate group II pre-miRNAs, which includes most of pre-let7 members, that shapes an optimal 3' end overhang for efficient processing (PubMed:25979828, PubMed:28671666). Add oligo-U tails to truncated pre-miRNAS with a 5' overhang which may promote rapid degradation of non-functional pre-miRNA species (PubMed:25979828). Does not play a role in replication-dependent histone mRNA degradation (PubMed:18172165). Due to functional redundancy between TUT4 and TUT7, the identification of the specific role of each of these proteins is difficult (PubMed:25979828, PubMed:25480299, PubMed:19703396, PubMed:22898984, PubMed:18172165, PubMed:28671666). TUT4 and TUT7 restrict retrotransposition of long interspersed element-1 (LINE-1) in cooperation with MOV10 counteracting the RNA chaperonne activity of L1RE1. TUT7 uridylates LINE-1 mRNAs in the cytoplasm which inhibits initiation of reverse transcription once in the nucleus, whereas uridylation by TUT4 destabilizes mRNAs in cytoplasmic ribonucleoprotein granules (PubMed:30122351). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ13409; PAP associated domain containing 6; Terminal uridylyltransferase 7; TUTase 7; TUTase7; Zinc finger CCHC domain-containing protein 6; zinc finger, CCHC domain containing 6
Gene Aliases: 6030448M23Rik; AA420405; HS2; KIAA1711; PAPD6; TUT7; ZCCHC6
UniProt ID: (Human) Q5VYS8, (Mouse) Q5BLK4
Entrez Gene ID: (Human) 79670, (Mouse) 214290, (Rat) 501515
Molecular Function:
nucleotidyltransferase
transferase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support