Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ZDHHC12 gene ontology annotations related to this gene include peptidyl-L-cysteine S-palmitoylation; protein palmitoylation; protein targeting to membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DHHC domain-containing cysteine-rich protein 12; DHHC-12; membrane-associated DHHC12 zinc finger protein; Palmitoyltransferase ZDHHC12; probable palmitoyltransferase ZDHHC12; Zinc finger DHHC domain-containing protein 12; Zinc finger protein 400; zinc finger, DHHC domain containing 12; zinc finger, DHHC-type containing 12
Gene Aliases: 1190004A01Rik; DHHC-12; PSEC0008; ZDHHC12; ZNF400
UniProt ID: (Human) Q96GR4, (Mouse) Q8VC90, (Rat) Q6DGF5
Entrez Gene ID: (Human) 84885, (Mouse) 66220, (Rat) 366014
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support