Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTLLTDATPL VKEPHPLPLV PRPWFLPSLF AAFNVVLLVF FSGLFFAFPL VSLNFSDPGI LHQGSAEQGP LTVHVVWVNH GAFRLQWCPK CCFHRPPRTY HCPWCNICVE VSAPLPVHQP SPHIWLGPGA SLIQGPGLGV KLFSGLLKAK GGSNSQPPLV THVFSTCTCP GRLAVACRGA DMHRASASVL GWVRPGEERQ PSVPPLGSLT RDTQAPQAGC PPGLGGLACP RLGVQVGGSP GSE
ZDHHC19 gene ontology annotations related to this gene include peptidyl-L-cysteine S-palmitoylation; protein targeting to membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DHHC-19; Palmitoyltransferase ZDHHC19; Zinc finger DHHC domain-containing protein 19; zinc finger, DHHC domain containing 19; zinc finger, DHHC-type containing 19
Gene Aliases: DHHC19; ZDHHC19
UniProt ID: (Human) Q8WVZ1
Entrez Gene ID: (Human) 131540
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support