Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YVLVLGPGPP PLGPLARALQ LALAAFQLLN LLGNVGLFLR SDPSIRGVML
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 93%
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DHHC-24; Probable palmitoyltransferase ZDHHC24; Zinc finger DHHC domain-containing protein 24; zinc finger, DHHC domain containing 24; zinc finger, DHHC-type containing 24
Gene Aliases: UNQ2528/PRO6027; ZDHHC24
UniProt ID: (Human) Q6UX98
Entrez Gene ID: (Human) 254359
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support