Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Dog, Orangutan and Chimpanzee
Palmitoyl acyltransferase for the G-protein coupled receptor SSTR5. Also palmitoylates FLOT2. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DHHC-5; membrane-associated DHHC5 zinc finger protein; Palmitoyltransferase ZDHHC5; probable palmitoyltransferase ZDHHC5; Zinc finger DHHC domain-containing protein 5; Zinc finger protein 375; zinc finger, DHHC domain containing 5; zinc finger, DHHC-type containing 5
Gene Aliases: 1110032A17Rik; AI451382; DHHC5; KIAA1748; ZDHHC5; Zisp; ZNF375
UniProt ID: (Human) Q9C0B5, (Mouse) Q8VDZ4
Entrez Gene ID: (Human) 25921, (Mouse) 228136
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support