Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: VEHVLCSPLA PRYVVEPPRL PLAVSLKPPF LRPELLDRAA PLKVKLSDNG LKAGLGRSKS KGSLDRLDEK PLDLGPPLPP KIEAGTFSSD LQTPRPGSAE SALSVQRTSP PTPAMYKFRP AFPTGPKVPF CGPGEQVPGP D
This gene encodes a four transmembrane protein that is a member of the zinc finger DHHC domain-containing protein family. The encoded protein may function as a palmitoyltransferase. Defects in this gene may be associated with a susceptibility to schizophrenia. Alternate splicing of this gene results in multiple transcript variants. A pseudogene of this gene is found on chromosome 22.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DHHC-8; DHHC-containing protein 8; membrane-associated DHHC8 zinc finger protein; Palmitoyltransferase ZDHHC8; probable palmitoyltransferase ZDHHC8; Zinc finger DHHC domain-containing protein 8; Zinc finger protein 378; zinc finger, DHHC domain like containing 1; zinc finger, DHHC-type containing 8
Gene Aliases: D16H22S1738E; DHHC-8; DHHC8; E330009O14Rik; KIAA1292; Op53c05; ZDHHC8; ZDHHCL1; ZNF378
UniProt ID: (Human) Q9ULC8, (Mouse) Q5Y5T5
Entrez Gene ID: (Human) 29801, (Mouse) 27801
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support