Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SFSDPGVIP RALPDEAAFI EMEIEATNGA VPQGQRPPPR IKNFQINNQI VKLKYCYTCK IFRPPRASHC SICDNCVERF DHHCPWVGNC VGKRNYRYFY LFILSLSLLT IYVFAFNIVY VALKSLKIGF LETLKETPGT VLEVLICFFT LWSVVGLTGF HTFLVALNQT TNEDIKGSWT GKNRVQNPYS HGNIVKNCCE VLCGPLPPSV LDRRGILPLE ESGSRPPSTQ ETSSSLLPQS PAPTEHLNSN EMPEDSSTPE EMPPPEPPEP PQEAAEAEK (87-364 aa encoded by BC006200)
The ZDHHC9-GOLGA7 complex is a palmitoyltransferase specific for HRAS and NRAS.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antigen MMSA-1; Asp-His-His-Cys domain containing protein 9; DHHC-9; DHHC9; Palmitoyltransferase ZDHHC9; Zinc finger DHHC domain-containing protein 9; Zinc finger protein 379; Zinc finger protein 380; zinc finger, DHHC domain containing 10; zinc finger, DHHC-type containing 9
Gene Aliases: 6430508G22; 9530098M12Rik; CGI-89; CGI89; CXorf11; DHHC9; MMSA1; MRXSZ; UNQ261/PRO298; ZDHHC10; ZDHHC9; ZNF379; ZNF380
UniProt ID: (Human) Q9Y397, (Mouse) P59268
Entrez Gene ID: (Human) 51114, (Mouse) 208884
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support