Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ZFAND3 (Zinc Finger AN1-Type Containing 3) is a Protein Coding gene and also known as Testis expressed sequence 27.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AN1-type zinc finger protein 3; RNF1; testis expressed sequence 27; Testis-expressed protein 27; zinc finger, AN1-type domain 3
Gene Aliases: TEX27; ZFAND3
UniProt ID: (Human) Q9H8U3
Entrez Gene ID: (Human) 60685
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support