Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
ZFP37 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zfp-37; Zinc finger protein 37 homolog; zinc finger protein homologous to Zfp37 in mouse
Gene Aliases: zfp-37; ZFP37; ZNF906
UniProt ID: (Human) Q9Y6Q3
Entrez Gene ID: (Human) 7539
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support