Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EEHQSPGGIS SEEEEEEEEE MLISEEEIPF KDDPRDETYK PHLERETPKP RRKSGKVKEE KEKKEIKVEV EVEVKEEENE IREDEE
Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.
ZFP91 is a member of the zinc finger family of proteins. The gene product contains C2H2 type domains, which are the classical zinc finger domains found in numerous nucleic acid-binding proteins. In addition to the monocistronic transcript originating from this locus, a co-transcribed variant composed of ZFP91 and CNTF sequence has been identified. The monocistronic and co-transcribed variants encode distinct isoforms. The co-transcription of ZFP91 and CNTF has also been observed in mouse.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: drug resistance-associated sequence in melanoma 8; E3 ubiquitin-protein ligase ZFP91; penta zinc finger protein; RING-type E3 ubiquitin transferase ZFP91; Zfp-91; Zinc finger protein 757; Zinc finger protein 91 homolog; zinc finger protein homologous to Zfp91 in mouse
Gene Aliases: DMS-8; DSM-8; FKSG11; PZF; ZFP-91; ZFP91; ZNF757
UniProt ID: (Human) Q96JP5
Entrez Gene ID: (Human) 80829
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support