Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: SDHRYRCQRG IVHITSESLA KDQLAIAASR HGRTYHAYPD FAPQKFKEKT QGQVKILEDS FLKSSFPTQA ELDRLRVETK LSRREIDSWF SERRKLRDSM EQAVLDSMGS GKKGQDVGAP NGALSRLDQL SGAQLTSSLP SPSPAIAKSQ EQVHLLRSTF ARTQWPTPQE YDQLAAKTGL VRTEIVRWFK ENRCLLKTGT VKWMEQYQHQ PMADDHGYDA VARKATKPMA ESPKNGGDVV PQYYKDPKKL CEEDLEKLVT RVKVGSEPAK DCLPAKPSEA TSDRSEGSSR DGQGSDENEE SSVVDYVEVT VGEEDAISDR SDSWSQAAAE GVSELAESDS DCVPAEAGQA
Zinc-fingers and homeobox (ZHX) proteins are transcription factors that interact with the activation domain of the A subunit of nuclear factor-Y (NF-YA). ZHX1-3 are ubiquitously expressed proteins expressed in various tissues. They act as transcriptional repressors and localize to the nucleus. The ZHX proteins contain two Cys(2)-His(2)-type zinc-finger motifs and five homeodomains (HDs). These domains allow the ZHX proteins to form homodimers, but they can also form heterodimers with each other. However, this dimerization is not required for repressor activity. Hypermethylation-mediated silencing of ZHX2 is an epigenetic event involved in hepatocellular carcinoma (HCC).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AFP regulator 1; alpha fetoprotein regulation 1, adult; Alpha-fetoprotein regulator 1; Regulator of AFP; transcription factor ZHX2; Zinc finger and homeodomain protein 2; Zinc fingers and homeoboxes protein 2; zinc-fingers and homeoboxes 2
Gene Aliases: Afr-1; AFR1; KIAA0854; mKIAA0854; RAF; ZHX2
UniProt ID: (Human) Q9Y6X8, (Rat) Q80VX4, (Mouse) Q8C0C0
Entrez Gene ID: (Human) 22882, (Pig) 100152952, (Rabbit) 100350637, (Rat) 314988, (Mouse) 387609
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support