Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 500 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in many species including rat and mouse.
Specificity of this antibody: ZIC1, 2, 3.
Zic1 encodes a zinc finger protein expressed in the developing or matured central nervous system in a highly restricted manner. Zic is expressed in granule cells that make synaptic contact with Purkinje cells. Zic1 is a gene critical to cerebellar pattern formation. The expression of Zic genes is first detected at gastrulation and at neurulation, becoming restricted to the dorsal neural ectoderm and the dorsal paraxial mesoderm. The Zic1 gene has been mapped to chromosome 9 in mouse. The 5' flanking region of the Zic1 gene contains a region-specific enhancer determined to be essential in in-vivo and in-vitro deletion analysis. The temporal profile of mRNA expression differs for each of the Zic gene products. The Drosophila odd-paired gene is highly homologous to the Zic gene family. Zic2 and Zic3 are highly similar genes. Zic2 is essential for the formation of the brain and Zic-3 is important for right and left axis formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GENA 29; heterotaxy 1; kumba; odd-paired homolog; Zic family member 1 (odd-paired homolog, Drosophila); Zic family member 2 (odd-paired homolog, Drosophila); Zic family member 3 (odd-paired homolog, Drosophila); zic protein member 1; Zinc finger protein 201; Zinc finger protein 203; Zinc finger protein of the cerebellum 1; Zinc finger protein of the cerebellum 2; Zinc finger protein of the cerebellum 3; Zinc finger protein ZIC 1; Zinc finger protein ZIC 2; Zinc finger protein ZIC 3
Gene Aliases: Bn; CRS6; HPE5; HTX; HTX1; Ka; Ku; RGD1561261; VACTERLX; ZIC; ZIC1; ZIC2; ZIC3; ZNF201; ZNF203
UniProt ID: (Human) Q15915, (Human) O95409, (Human) O60481, (Mouse) P46684, (Mouse) Q62521
Entrez Gene ID: (Human) 7545, (Human) 7546, (Human) 7547, (Rat) 64618, (Mouse) 22771, (Rat) 361096, (Mouse) 22772, (Mouse) 22773, (Rat) 367944
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support