Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-31710 has been tested in indirect ELISA.
DAPK3 is a serine/threonine kinase which is functionally linked to cell motility, smooth muscle contractility and cell death processes. DAPK3 is encoded by ZIPK and is a component of desmosomes and of the epidermal cornified envelope in keratinocytes. The N-terminal domain of this protein interacts with the plasma membrane and its C-terminus interacts with intermediate filaments. Through its rod domain, this protein forms complexes with envoplakin. This protein may serve as a link between the cornified envelope and desmosomes as well as intermediate filaments. AKT1/PKB, a protein kinase mediating a variety of cell growth and survival signaling processes, is reported to interact with this protein, suggesting a possible role for this protein as a localization signal in AKT1-mediated signaling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAP kinase 3; DAP-like kinase; DAP3K; Death-associated protein kinase 3; Dlk; MYPT1 kinase; ZIP kinase isoform; ZIP-kinase; Zipper-interacting protein kinase
Gene Aliases: DAPK3; DLK; ZIP; ZIPK
UniProt ID: (Human) O43293
Entrez Gene ID: (Human) 1613
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support