Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
ZKSCAN4 may be involved in the transcriptional activation of MDM2 and EP300 genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: P373c6.1; p373c6.1 (novel C2H2 type zinc finger protein); Zinc finger protein 307; Zinc finger protein 427; Zinc finger protein with KRAB and SCAN domains 4
Gene Aliases: P1P373C6; ZKSCAN4; ZNF307; ZNF427; ZSCAN36
UniProt ID: (Human) Q969J2
Entrez Gene ID: (Human) 387032
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support