Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody detects endogenous levels of total ZMIZ1 protein.
Retinoic acid plays a critical role in development, cellular growth, and differentiation and induces the expression of a variety of genes. RAI17 expression is induced by retinoic acid and is predominantly expressed in heart, brain and ovaries. Within brain, highest expression is in amygdala. The deduced 1,067-amino acid protein contains an MSX-interacting zinc finger (MIZ) domain, a nuclear localization signal sequence, and 2 proline-rich regions. A strong intrinsic transactivation domain is identified in the C-terminal proline-rich region. RAI17 expression is detected in various cancer cell lines. RAI17 colocalizes with endogenous androgen receptor (AR) in the nuclei of prostate epithelial cells from human tissue samples. In human prostate cancer cells, RAI17 increases the transcriptional activity of AR. Studies using sumoylation-deficient AR mutants suggest that the increase of AR activity by RAI17 is dependent upon receptor sumoylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ00092 protein; PIAS-like protein Zimp10; retinoic acid induced 17; Retinoic acid-induced protein 17; RP11-519K18.1; Zinc finger MIZ domain-containing protein 1; zinc finger, MIZ-type containing 1; zinc finger-containing, Miz1, PIAS-like protein on chromosome 10
Gene Aliases: BC065120; E330020C23; ENSMUSG00000072684; Gm10397; hZIMP10; I920194n01; KIAA1224; MIZ; RAI17; TRAFIP10; ZIMP10; ZMIZ1
UniProt ID: (Human) Q9ULJ6, (Mouse) Q6P1E1
Entrez Gene ID: (Human) 57178, (Mouse) 328365
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support