Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: GEKPYECKQC GKAFSYLNSF RTHEMIHTGE KPFECKRCGK AFRSSSSFRL HERTHTGQKP YHCKECGKAY SCRASFQRHM LTHAEDGPPY KCMWESL
ZNF136 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 14 C2H2-type zinc fingers and 1 KRAB domain. ZNF136 may be involved in transcriptional regulation as a weak repressor when alone, or a potent one when fused with a heterologous protein containing a KRAB B-domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 136
Gene Aliases: pHZ-20; ZNF136
UniProt ID: (Human) P52737
Entrez Gene ID: (Human) 7695
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support