Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ZNF148 is involved in transcriptional regulation. ZNF148 represses the transcription of a number of genes including gastrin, stromelysin and enolase. ZNF148 binds to the G-rich box in the enhancer region of these genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta enolase repressor factor 1; CACCC box-binding protein; CLL-associated antigen KW-10; G-rich box-binding protein; Transcription factor BFCOL1; Transcription factor ZBP-89; Zinc finger DNA-binding protein 89; Zinc finger protein 148
Gene Aliases: 2210405J08Rik; AI480666; AW045217; BERF-1; BFCOL1; HT-BETA; pHZ-52; ZBP-89; ZBP89; ZFP148; ZNF148
UniProt ID: (Human) Q9UQR1, (Rat) Q62806, (Mouse) Q61624
Entrez Gene ID: (Human) 7707, (Rat) 58820, (Mouse) 22661
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support