Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: KEKEPRVEEA EVSHQRCQER EFGLEIPQKE ISKKASFQKD MVGEFTRDGS WCSILEELRL DADRTKKDEQ NQIQPMSHSA FFNKKTLNTE SNCEYKDP
ZNF175 expression in brain mononuclear phagocytes is a signature for advanced HIV-1 encephalitis. As a transcriptional suppressor, this protein plays a role in macrophage control of viral replication during advanced HIV-1 infection.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 175; Zinc finger protein OTK18
Gene Aliases: OTK18; ZNF175
UniProt ID: (Human) Q9Y473
Entrez Gene ID: (Human) 7728
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support