Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
Recommended positive controls: A549, HeLa, HepG2, HCT116.
Predicted reactivity: Xenopus laevis (81%), Rhesus Monkey (100%), Chimpanzee (83%), Bovine (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ZNF207 is a new candidate transcription factor.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BUB3-interacting and GLEBS motif-containing protein ZNF207; BuGZ; Zinc finger protein 207
Gene Aliases: BUGZ; hBuGZ; ZNF207
UniProt ID: (Human) O43670
Entrez Gene ID: (Human) 7756
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support