Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MGRKKKKQLK PWCWYCNRDF DDEKILIQHQ KAKHFKCHIC HKKLYTGPGL AIHCMQVHKE TIDAVPNAIP GRTDIELEIY GMEGIPEKDM DERRRLLEQK TQESQKKKQQ
ZNF207 is a new candidate transcription factor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BUB3-interacting and GLEBS motif-containing protein ZNF207; BuGZ; Zinc finger protein 207
Gene Aliases: BUGZ; hBuGZ; ZNF207
UniProt ID: (Human) O43670
Entrez Gene ID: (Human) 7756
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support