Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LQHPNNSKD MVTLIEDVIE MLEDEDMPCK DSALQMGSIK EKMKAGSRTG KPQEPVTFKD VVVEFSKEEW GQLDSAVKNL YRNVMLENFR NLNSLRKAHL LSKPFESLKL ESKKKRWIME KEIPRKTIFD MKSISGEESS HGVIMTRLTE SGHPSSDAWK DGVSLLLPRL EYNDAVSAHC NLHLWVQVIL LPQPPE (110-304 aa encoded by BC014538)
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BAZ 2; BAZ-2; BWSCR2-associated zinc finger protein 2; Zinc finger protein 215; Zinc finger protein with KRAB and SCAN domains 11
Gene Aliases: BAZ-2; BAZ2; ZKSCAN11; ZNF215; ZSCAN43
UniProt ID: (Human) Q9UL58
Entrez Gene ID: (Human) 7762
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support