Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MEFPDHSRHL LQCLSEQRHQ GFLCDCTVLV GDAQFRAHRA VLASCSMYFH LFYKDQLDKR DIVHLNSDIV TAPAFALLLE FMYEGKLQFK DLPIEDVLAA ASYLHMYDIV KVCKKKLKEK ATTEADSTKK EEDASSCSDK VESLSDGSSH IAGDLPSDED EGEDEKLNIL PSKRDLAAEP GNMWMRLPSD SAGIPQAGGE AEPHATAAGK TVASPCSSTE SLSQRSVTSV RDSADVDCVL DLSVKSSLSG VENLNSSYFS SQDVLRSNLV QVKVEKEASC DESDVGTNDY DMEHSTVKES VSTNNRVQYE PAHLAPLRED SVLRELDRED KASDDEMMTP ESERVQVEGG MESSLLPYVS NILSPAGQIF MCPLCNKVFP SPHILQIHLS THFREQDGIR SKPAADVNVP TCSLCGKTFS CMYTLKRHER THSGEKPYTC TQCGKSFQYS HNLSRHAVVH TREKPHACKW CERRFTQSGD LYRHIRKFHC ELVNSLSVKS EALSLPTVRD WTLEDSSQEL WK
ZNF238 is the sequence-specific DNA-binding protein with transcriptional repression activity. ZNF238 binds the consensus DNA sequence 5'-[AC]ACATCTG[GT][AC]-3' which contains the E box core. ZNF238 may play a role in the organization of chromosomes in the nucleus.C2H2-type zinc finger proteins, such as ZNF238, act on the molecular level as transcriptional activators or repressors and are involved in chromatin assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 58 kDa repressor protein; TAZ-1; Transcriptional repressor RP58; Translin-associated zinc finger protein 1; Zinc finger and BTB domain-containing protein 18; Zinc finger protein 238; Zinc finger protein C2H2-171
Gene Aliases: C2H2-171; MRD22; RP58; TAZ-1; TAZ1; ZBTB18; ZNF238
UniProt ID: (Human) Q99592
Entrez Gene ID: (Human) 10472
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support