Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: QKIDNPESQA NSGALDTNQV LLHKIPPRKR LRKRDSQVKS MKHNSRVKIH QKSCERQKAK EGNGCRKTFS RSTKQITFIR IHKGSQVCRC SECGKIFRNP RYFSVHKKIH T
ZNF274 is a zinc finger protein containing five C2H2-type zinc finger domains, one or two Kruppel-associated box A (KRAB A) domains, and a leucine-rich domain. The protein has been suggested to be a transcriptional repressor. It localizes predominantly to the nucleolus.This gene encodes a zinc finger protein containing five C2H2-type zinc finger domains, one or two Kruppel-associated box A (KRAB A) domains, and a leucine-rich domain. The encoded protein has been suggested to be a transcriptional repressor. It localizes predominantly to the nucleolus. Alternatively spliced transcript variants encoding different isoforms exist. These variants utilize alternative polyadenylation signals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686K08243; KRAB zinc finger protein HFB101; Neurotrophin receptor-interacting factor homolog; Zf2; Zinc finger protein 274; Zinc finger protein HFB101; Zinc finger protein with KRAB and SCAN domains 19; Zinc finger protein zfp2
Gene Aliases: HFB101; SP2114; ZF2; ZKSCAN19; ZNF274; ZSCAN51
UniProt ID: (Human) Q96GC6
Entrez Gene ID: (Human) 10782
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support