Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nucleolar autoantigen 36; Nucleolar cysteine-rich protein; zinc finger autoantigen 330; Zinc finger protein 330
Gene Aliases: HSA6591; NOA36; ZNF330
UniProt ID: (Human) Q9Y3S2
Entrez Gene ID: (Human) 27309
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support