Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAEGSVMFSD VSIDFSQEEW DCLDPVQRDL YRDVMLENYG NLVSMGLYTP KPQVISLLEQ GKEPWMVGRE LTRGLCSDLE SMCETKLLSL KKEVYEIELC QREIMGLTKH GLEYSSFGDV LEYRSHLAKQ LGYPNGHFSQ EIFTPEYMPT FIQQTFLTLH QIINNEDRPY ECKKCGKAFS QNSQFIQHQR IHIGEKSYEC KECGKFFSCG SHVTRHLKIH TGEKPFECKE CGKAFSCSSY LSQHQRIHTG KKPYECKECG KAFSYCSNLI DHQRIHTGEK PYECKVCGKA FTKSSQLFQH ARIHTGEKPY ECKECGKAFT QSSKLVQHQR IHTGEKPYEC KECGKAFSSG SALTNHQRIH TGEKPYDCKE CGKAFTQSSQ LRQHQRIHAG EKPFECLECG KAFTQNSQLF QHQRIHTDEK PYECNECGKA FNKCSNLTRH LRIHTGEKPY NCKECGKAFS SGSDLIRHQG IHTNK
May function as a transcriptional repressor, suppressing transcriptional activities mediated by MAPK signaling pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 383
Gene Aliases: HSD17; ZNF383
UniProt ID: (Human) Q8NA42
Entrez Gene ID: (Human) 163087
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support