Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PVSMENGLGP APGSPEKQPG SPSPPSIPET GQGVTKGEGG TPAPASLPGG SKEEE
Antigen sequence identity: Mouse 93%; Rat 93%
ZNF385 is a new candidate transcription factor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hematopoietic zinc finger protein; Retinal zinc finger protein; zinc finger protein 385; Zinc finger protein 385A
Gene Aliases: HZF; RZF; ZFP385; ZNF385; ZNF385A
UniProt ID: (Human) Q96PM9
Entrez Gene ID: (Human) 25946
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support