Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
ZNF394 belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SNAP19; truncated zinc finger protein isoform; Zinc finger protein 394; zinc finger protein 99; Zinc finger protein with KRAB and SCAN domains 14
Gene Aliases: ZKSCAN14; ZNF394; ZSCAN46
UniProt ID: (Human) Q53GI3
Entrez Gene ID: (Human) 84124
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support