Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NYSCGKRTKA FSTKHSVIPH QKLFT
Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 48%.
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 417
Gene Aliases: ZNF417
UniProt ID: (Human) Q8TAU3
Entrez Gene ID: (Human) 147687
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support