Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSTAPSLSAL RSSKHSGGGG GGGGGGGADP AWTSALSGNS SGPGPGSSPA GSTKPFVHAV PPSDPLRQAN RLPIKVLKML TARTGHILHP EYLQPLPSTP VSPIELDAKK SPLALLAQTC SQIGKPDPSP SSKLSSVASN GGGAGGAGGG AAGDKDTKSG PLKLSDIGVE DKSSFKPYSK PGSDKKEPGG GGGGGGGGGG GGGGVSSEKS GFRVPSATCQ PFTPRTGSPS SSASACSPGG MLSSAGGAPE GKDDKKDTDV GGGGKGTGGA SAEGGPTGLA HGRISCGGGI NVDVNQHPDG GPGGKALGSD CGGSSGSSSG SGPSAPTSSS VLGSGLVAPV SPYKPGQTVF PLPPAGMTYP GSLAGAYAGY PPQFLPHGVA LDPTKPGSLV GAQLAAAAAG SLGCSKPAGS SPLAGASPPS VMTASLCRDP YCLSYHCASH LAGAAAASAS CAHDPAAAAA ALKSGYPLVY PTHPLHGVHS SLTAAAAAGA TPPSLAGHPL YPYGFMLPND PLPHICNWVS ANGPCDKRFA TSEELLSHLR THTAFPGTDK LLSGYPSSSS LASAAAAAMA CHMHIPTSGA PGSPGTLALR SPHHALGLSS RYHPYSKSPL PTPGAPVPVP AATGPYYSPY ALYGQRLTTA SALGYQ
May function as a transcriptional repressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 503
Gene Aliases: Nlz2; NOLZ-1; NOLZ1; ZNF503
UniProt ID: (Human) Q96F45
Entrez Gene ID: (Human) 84858
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support