Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: SYHHSFQSRG RPHTGKKRYE CKECGKTFSS RRNLRRHMVV QGGNRPYKFP
Sequence homology: Human: 100%
ZNF563 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 563
Gene Aliases: ZNF563
UniProt ID: (Human) Q8TA94
Entrez Gene ID: (Human) 147837
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support