Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ZNF622 may behave as an activator of the bound transcription factor, MYBL2, and be involved in embryonic development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytoplasmic 60S subunit biogenesis factor ZNF622; Zinc finger protein 622
Gene Aliases: 1110033B05Rik; D15Ertd806e; Zfp622; Znf622; ZPR9
UniProt ID: (Mouse) Q91VY9
Entrez Gene ID: (Mouse) 52521
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support