Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SGLITQEGVH IPFDVHHVES LAEQGTPLCS NPAGNGPEAL ETVVCVPVPV QVGAGPSALF ENVPQEALGE VVASCPMPGM VPGSQVIIIA GPGYDALTAE GIHLNMAAGS GVPGSG
Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 67 kDa zinc finger protein; Zinc finger protein 653; Zinc finger protein Zip67
Gene Aliases: E430039K05Rik; ZIP67; ZNF653
UniProt ID: (Human) Q96CK0
Entrez Gene ID: (Human) 115950
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support