Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAPPPAPLLA PRPGETRPGC RKPGTVSFAD VAVYFSPEEW GCLRPAQRAL YRDVMQETYG HLGALGFPGP KPALISWMEQ ESEAWSPAAQ DPEKGERLGG ARRGDVPNRK EEEPEEVPRA KGPRKAPVKE SPEVLVERNP DPAISVAPAR AQPPKNAAWD PTTGAQPPAP IPSMDAQAGQ RRHVCTDCGR RFTYPSLLVS HRRMHSGERP FPCPECGMRF KRKFAVEAHQ WIHRSCSGGR RGRRPGIRAV PRAPVRGDRD PPVLFRHYPD IFEECG
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 688
Gene Aliases: ZNF688
UniProt ID: (Human) P0C7X2
Entrez Gene ID: (Human) 146542
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support