Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LISWLERNT DDWEPAALDP QEYPRGLTVQ RKSRTRKKNG EKEVFPPKEA PRKGKRGRRP SKPRLIPRQT SGGPICPDCG CTFPDHQALE SHKCAQNL (77-173 aa encoded by BC014000 )
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: transcription-involved protein upregulated in HCC 1; Zinc finger protein 689
Gene Aliases: TIPUH1; ZNF689
UniProt ID: (Human) Q96CS4
Entrez Gene ID: (Human) 115509
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support