Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 11
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hZNF3; Zinc finger protein 69; ZNF3
Gene Aliases: Cos5; hZNF3; ZNF69
UniProt ID: (Human) Q9UC07
Entrez Gene ID: (Human) 7620
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support