Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGSEKEQSPE PHLPEEGEGG KPWRVDDSEG SWIPPGEKEH GQESLSDELQ ETHPKKPWQK VTVRARELGD PIAHPRHEAD EKPFICAQCG KTFNNTSNLR THQRIHTGEK PYKCSECGKS FSRSSNRIRH ERIHLEEKHY KCPKCQESFR RRSDLTTHQQ DHLGKRPYRC DICGKSFSQS ATLAVHHRTH LEPAPYICCE CGKSFSNSSS FGVHHRTHTG ERPYECTECG RTFSDISNFG AHQRTHRGEK PYRCTVCGKH FSRSSNLIRH QKTHLGEQAG KDSS
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RP11-342M1.5; Zinc finger protein 691
Gene Aliases: Zfp691; ZNF691
UniProt ID: (Human) Q5VV52
Entrez Gene ID: (Human) 51058
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support