Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The calculated molecular weight of ZNF746 is 69 kDa, but phosphorylated ZNF746 is about 80-85 kDa.
Immunogen sequence: MQIKQEGEL QLQEQQALGV EAWAAGQPDI GEEPWGLSQL DSGAGDISTD ATSGVHSNFS TTIPPTSWQT DLPPHHPSSA CSDGTLKLNT AASTEDVKIV IKTEVQEEEV VATPVHPTDL EAHGTLFGPG QATRFFPSPA QEGAWESQGS SFPSQDPVLG LREPARPERD MGELSPAVAQ EETPPGDWLF GGVRWGWNFR CKPPVGLNPR TGPEGLPYSS PDNGEAILDP SQAPRPFNEP CKYPGRTKGF GHKPGLKKHP AAPPGGRPFT CATCGKSFQL QVSLSAHQRS CGAPDGSGPG TGGGGSGSGG GGGGSGGGSA RDGSALRCGE CGRCFTRPAH LIRHRML (186-531 aa encoded by BC068505 )
ZNF746 is a transcription repressor that specifically binds to the 5'-TATTTT[T/G]-3' consensus sequence on promoters and represses transcription of PGC-1-alpha (PPARGC1A), thereby playing a role in regulation of neuron death.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PARIS; parkin interacting substrate; Parkin-interacting substrate; parkin-interacting sustrate; Zinc finger protein 746
Gene Aliases: 2810407L07Rik; AI317225; PARIS; RGD1306209; Zfp746; ZNF746
UniProt ID: (Human) Q6NUN9, (Mouse) Q3U133
Entrez Gene ID: (Human) 155061, (Mouse) 69228, (Rat) 312303
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support