Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SSSFQAHKR THTGGKPYEC KQCGKSFSWC HSFQIHERTH TGEKPCECSK CNKAFRSYRS YLRHKRSHTG EKPYQCKECR KAFTYPSSLR RHERTHSAKK PYECKQCGKA LSYKFSNTPK NALWRKTL (268-394 aa encoded by BC105713)
ZNF763 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-binding protein; zinc finger protein 440 like; Zinc finger protein 763
Gene Aliases: ZNF; ZNF440L; ZNF763
UniProt ID: (Human) Q0D2J5
Entrez Gene ID: (Human) 284390
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support