Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAAEPMGPA QVPMNSEVIV DPIQGQVNFE DVFVYFSQEE WVLLDEAQRL LYRDVMLENF ALMASLGHTS FMSHIVASLV MGSEPWVPDW VDMTLAVATE TPGGSDPGCW HGMEDEEIPF EQSFSIGMSQ IRIPKGGPST QKAYPCGTCG LVLKDILHLA EHQETHPGQK PYMCVLCGKQ FCFSANLHQH QKQHSGEKPF RSDKSRPFLL NNCAVQSMEM SFVTGEACKD FLASSSIFEH HAPHNEWKPH SNTKCEEASH CGKRHYKCSE CGKTFSRKDS LVQHQRVHTG ERPYECGECG KTFSRKPILA QHQRIHTGEM PYECGICGKV FNHSSNLIVH QRVHTGARPY KCSECGKAYS HKSTLVQHES IHTGERPYEC SECGKYFGHK YRLIKHWSVH TGARPYECIA CGKFFSQSSD LIAHQRVHNG EKPYVCSECG KAFSHKHVLV QHHRIHTGER PYKCSECGKA FRQRASLIRH WKIHTGERP
ZNF772 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 772
Gene Aliases: ZNF772
UniProt ID: (Human) Q68DY9
Entrez Gene ID: (Human) 400720
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support