Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: GPLTFRDVAI EFSLKEWQCL DTAQRNLYRN VMLENYRNLV FLGITVSKPD LITCLEQGKE AWSMKRHEIM VAKPT
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 85; zinc finger protein 85 (HPF4, HTF1); Zinc finger protein HPF4; Zinc finger protein HTF1
Gene Aliases: HPF4; HTF1; ZNF85
UniProt ID: (Human) Q03923
Entrez Gene ID: (Human) 7639
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support