Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: WNVLMDYCYT TPSGNPNDDI RYDLFLSCDK DPQTTVIENG RSQRGRFSF EVFRFVKHKN QKMSTVFLHCV TKLCRADDCP FLMPICSHRE RRDAGRRTTW
ZPLD1 is a protein coding gene. Glycoprotein which is a component of the gelatinous extracellular matrix in the cupulae of the vestibular organ.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cupulin; zona pellucida-like domain containing 1; Zona pellucida-like domain-containing protein 1; ZP domain-containing protein 1
Gene Aliases: ZPLD1
UniProt ID: (Human) Q8TCW7
Entrez Gene ID: (Human) 131368
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support