Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: ITKQQTKQNC TKRYITKEDV AVASMDKVKN VGGHVRLITK ESRPRDPFTK KLLEDGACVP FLNPYTVQAD LTVKPSTSKG YLQAVDNEGN PLCLRCQQPT CQTKQACKAN SWDSRFCSLK CQEEFWIRSN NSYLRAKVFE TEHGVCQLCN VNAQELFLRL RDAPKSQRKN LLYATWTSKL PLEQLNEMIR NPGEGHFWQV DHIKPVYGGG GQCSLDNLQT LCTVCHKERT ARQAKERSQV RRQSLASKHG SDITRFLVKK; Positive Samples: HeLa, Raji, Jurkat, 293T, Mouse liver, Mouse heart, Mouse brain, Rat liver; Cellular Location: Chromosome, Nucleus
DNA annealing helicase and endonuclease required to maintain genome stability at stalled or collapsed replication forks by facilitating fork restart and limiting inappropriate recombination that could occur during template switching events (PubMed:21078962, PubMed:22704558, PubMed:22705370, PubMed:22759634, PubMed:26884333). Recruited to the sites of stalled DNA replication by polyubiquitinated PCNA and acts as a structure-specific endonuclease that cleaves the replication fork D-loop intermediate, generating an accessible 3'-OH group in the template of the leading strand, which is amenable to extension by DNA polymerase (PubMed:22759634). In addition to endonuclease activity, also catalyzes the fork regression via annealing helicase activity in order to prevent disintegration of the replication fork and the formation of double-strand breaks (PubMed:22705370, PubMed:22704558). [UniProt]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AH2; Annealing helicase 2; DNA annealing helicase and endonuclease ZRANB3; Zinc finger Ran-binding domain-containing protein 3; zinc finger, RAN-binding domain containing 3
Gene Aliases: 4933425L19Rik; AH2; AI316834; C730006D09; RGD1564952; ZRANB3
UniProt ID: (Human) Q5FWF4, (Mouse) Q6NZP1
Entrez Gene ID: (Human) 84083, (Rat) 304761, (Mouse) 226409
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support