Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
May be involved in transcriptional regulation during the post-meiotic stages of spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zfp-29; Zinc finger and SCAN domain-containing protein 2; Zinc finger protein 29 homolog; Zinc finger protein 854
Gene Aliases: ZFP29; ZNF854; ZSCAN2
UniProt ID: (Human) Q7Z7L9
Entrez Gene ID: (Human) 54993
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support