Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: masfvtevla hsgrleked l gtrisrlt rr veeikge vcn miskky sefl psmqs aqgli tqvd klsedi dll ksriese vr rdlhvstg e ftdlkqqle
This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Centromere/kinetochore protein zw10 homolog; H KNTC1AP; MGC149821; zeste white 10 homolog; ZW10 homolog, centromere/kinetochore protein; ZW10, kinetochore associated, homolog
Gene Aliases: HZW10; KNTC1AP; ZW10
UniProt ID: (Human) O43264
Entrez Gene ID: (Human) 9183
Molecular Function: microtubule or microtubule-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support